Assay Details
| Gene Symbol: Smndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172429.2 | NP_766017.1 | 3 - 4 | 423 | 71 | |
| GenBank mRNA | AK077225.1 | - | 3 - 4 | 445 | 71 | |
| AK078834.1 | - | 3 - 4 | 423 | 71 | ||
| AK090047.1 | - | 3 - 4 | 429 | 71 | ||
| BC058779.1 | - | 3 - 4 | 404 | 6312831 | 71 | |
| BC098223.1 | - | 2 - 3 | 286 | 1282458 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Smndc1
Genomic Map