Assay Details
| Gene Symbol: Smndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172429.2 | NP_766017.1 | 4 - 5 | 576 | 73 | |
| GenBank mRNA | AK010395.1 | - | 4 - 5 | 557 | 73 | |
| AK077225.1 | - | 4 - 5 | 598 | 73 | ||
| AK078834.1 | - | 4 - 5 | 576 | 73 | ||
| AK090047.1 | - | 4 - 5 | 582 | 73 | ||
| AK168575.1 | - | 4 - 5 | 550 | 73 | ||
| BC058779.1 | - | 4 - 5 | 557 | 6312831 | 73 | |
| BC098223.1 | - | 3 - 4 | 439 | 1282458 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Smndc1
Genomic Map