Assay Details
| Gene Symbol: Anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085383.1 | NP_001078852.1 | 1 - 2 | 123 | 82 | |
| GenBank mRNA | AK003395.1 | - | 1 - 2 | 123 | 82 | |
| BC062140.1 | - | 1 - 2 | 113 | 30290956 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003395.1, BC062140.1, NM_001085383.1 transcripts.
Set Membership: |
|

Anxa9
Genomic Map