Assay Details
| Gene Symbol: Fam204a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029648.6 | NP_083924.2 | 6 - 7 | 1020 | 69 | |
| GenBank mRNA | AK011404.2 | - | 5 - 6 | 911 | 69 | |
| AK075914.1 | - | 6 - 7 | 1021 | 69 | ||
| AK162079.1 | - | 6 - 7 | 972 | 69 | ||
| BC048505.1 | - | 6 - 7 | 559 | 69 | ||
| BC056640.1 | - | 2 - 3 | 69 | |||
| BC061459.1 | - | 819 | 6829614 | 69 | ||
More Information
Set Membership: |
|

Fam204a
Genomic Map