Assay Details
| Gene Symbol: Rab11fip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033172.3 | NP_001028344.2 | 2 - 3 | 1157 | 76 | |
| NM_001164367.1 | NP_001157839.1 | 2 - 3 | 585 | 76 | ||
| GenBank mRNA | AK015534.1 | - | 2 - 3 | 586 | 76 | |
| AK154916.1 | - | 2 - 3 | 1157 | 76 | ||
| BC089010.1 | - | 1 - 2 | 416 | 76 | ||
| BC139380.1 | - | 2 - 3 | 920 | 8862402 | 76 | |
| BC139419.1 | - | 2 - 3 | 920 | 8862441 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip2
Genomic Map