Assay Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB049956.1 | - | 1 - 2 | 33 | 72 | |
| AK002588.1 | - | 1 - 2 | 36 | 72 | ||
| AK007662.1 | - | 1 - 2 | 37 | 72 | ||
| AK008265.1 | - | 1 - 2 | 51 | 72 | ||
| AK009703.1 | - | 1 - 2 | 37 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AB049956.1,AK002588.1,AK007662.1,AK008265.1,AK009703.1 transcripts.
This assay is in the 5'UTR region of the AB049956.1,AK002588.1,AK007662.1,AK008265.1,AK009703.1 transcripts.
Set Membership: |
|

Mrps21
Genomic Map