Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800009972.mm.1 |
NM_001127338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009972.mm.1 |
NM_001127338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009972.mm.1 |
NM_138600.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009972.mm.1 |
NM_138600.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009972.mm.1 |
TC1800001366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800009974.mm.1 |
NM_001127338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009974.mm.1 |
NM_001127338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009974.mm.1 |
NM_138600.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009974.mm.1 |
NM_138600.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009974.mm.1 |
TC1800001366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800009973.mm.1 |
NM_001127338.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009973.mm.1 |
NM_001127338.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009973.mm.1 |
NM_138600.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009973.mm.1 |
NM_138600.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800009973.mm.1 |
TC1800001366.mm.1 |
MTA TC |