Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293686.1 | NP_001280615.1 | 4 - 5 | 737 | 86 | |
| NM_016862.4 | NP_058558.1 | 4 - 5 | 737 | 86 | ||
| GenBank mRNA | AF035209.1 | - | 4 - 5 | 444 | 86 | |
| AF035823.1 | - | 4 - 5 | 442 | 86 | ||
| AK004751.1 | - | 4 - 5 | 433 | 86 | ||
| AK028646.1 | - | 4 - 5 | 443 | 86 | ||
| AK135463.1 | - | 4 - 5 | 500 | 86 | ||
| AK148281.1 | - | 4 - 5 | 468 | 86 | ||
| BC012211.1 | - | 4 - 5 | 439 | 86 | ||
| BC019386.1 | - | 4 - 5 | 739 | 5012235 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vti1a
Genomic Map