Assay Details
| Gene Symbol: Akr1cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027582.3 | NP_081858.2 | 1 - 2 | 163 | 79 | |
| GenBank mRNA | AK087842.1 | - | 1 - 2 | 164 | 79 | |
| AK135492.1 | - | 1 - 2 | 166 | 79 | ||
| BC028632.1 | - | 1 - 2 | 147 | 79 | ||
| BC051128.1 | - | 1 - 2 | 163 | 6707386 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0900002122.rn
Additional Information:
This assay is in the 5'UTR region of the AK135492.1,BC051128.1,NM_027582.3 transcripts.
Set Membership: |
|

Akr1cl
Genomic Map