Assay Details
| Gene Symbol: Fzd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042659.1 | NP_001036124.1 | 1 - 2 | 514 | 60 | |
| GenBank mRNA | AK039402.1 | - | 1 - 2 | 513 | 60 | |
| BC117723.2 | - | 1 - 2 | 47 | 40088671 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC117723.2, NM_001042659.1 transcripts.
Set Membership: |
|

Fzd5
Genomic Map