Assay Details
| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037726.1 | NP_001032815.1 | 2 - 3 | 360 | 100 | |
| NM_133828.2 | NP_598589.2 | 3 - 4 | 482 | 100 | ||
| GenBank mRNA | AK042595.1 | - | 3 - 4 | 455 | 100 | |
| AK044992.1 | - | 3 - 4 | 297 | 100 | ||
| AK044993.1 | - | 3 - 4 | 297 | 100 | ||
| AK079223.1 | - | 3 - 4 | 478 | 100 | ||
| U46027.1 | - | 2 - 3 | 230 | 100 | ||
| X92497.1 | - | 2 - 3 | 337 | 100 | ||
More Information
Set Membership: |
|

Creb1
Genomic Map