Assay Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016878.4 | NP_058574.3 | 1 - 2 | 126 | 57 | |
| GenBank mRNA | AK052001.1 | - | 104 | 57 | ||
| AK166276.1 | - | 1 - 2 | 76 | 57 | ||
| BC092232.1 | - | 1 - 2 | 54 | 30551654 | 57 | |
| BY197391.1 | - | 1 - 2 | 126 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052001.1, AK166276.1, BC092232.1, NM_016878.4 transcripts.
Set Membership: |
|

Dnpep
Genomic Map