Assay Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205236.1 | NP_001192165.1 | 17 - 18 | 3003 | 56 | |
| NM_133715.5 | NP_598476.2 | 15 - 16 | 1972 | 56 | ||
| GenBank mRNA | AK046763.1 | - | 15 - 16 | 1973 | 56 | |
| AK143916.1 | - | 13 - 14 | 1444 | 56 | ||
| AK154926.1 | - | 16 - 17 | 3294 | 56 | ||
| BC096671.1 | - | 2 - 3 | 198 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap27
Genomic Map