Assay Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205236.1 | NP_001192165.1 | 5 - 6 | 1827 | 57 | |
| NM_133715.5 | NP_598476.2 | 3 - 4 | 796 | 57 | ||
| GenBank mRNA | AK046763.1 | - | 3 - 4 | 797 | 57 | |
| AK143916.1 | - | 1 - 2 | 268 | 57 | ||
| AK154926.1 | - | 5 - 6 | 1828 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap27
Genomic Map