Assay Details
| Gene Symbol: Slc6a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201353.1 | NP_958741.1 | 7 - 8 | 1154 | 66 | |
| GenBank mRNA | AK141581.1 | - | 7 - 8 | 1179 | 66 | |
| BC050103.1 | - | 7 - 8 | 1055 | 30024098 | 66 | |
| BC057070.1 | - | 7 - 8 | 1154 | 5695483 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a7
Genomic Map