Assay Details
| Gene Symbol: Slc6a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201353.1 | NP_958741.1 | 10 - 11 | 1520 | 75 | |
| GenBank mRNA | AK141581.1 | - | 10 - 11 | 1545 | 75 | |
| BC050103.1 | - | 10 - 11 | 1421 | 30024098 | 75 | |
| BC057070.1 | - | 10 - 11 | 1520 | 5695483 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a7
Genomic Map