Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175751.4 | NP_786927.2 | 2 - 3 | 1448 | 61 | |
| GenBank mRNA | AK038794.1 | - | 1 - 2 | 86 | 61 | |
| AK047979.1 | - | 2 - 3 | 1409 | 61 | ||
| BC092219.1 | - | 2 - 3 | 1181 | 6830664 | 61 | |
| BC151073.1 | - | 1 - 2 | 967 | 9087986 | 61 | |
| BC151075.1 | - | 1 - 2 | 967 | 9087988 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp608
Genomic Map