Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175751.4 | NP_786927.2 | 3 - 4 | 1707 | 65 | |
| GenBank mRNA | AK038794.1 | - | 2 - 3 | 345 | 65 | |
| AK077030.1 | - | 2 - 3 | 431 | 65 | ||
| BC092219.1 | - | 3 - 4 | 1440 | 6830664 | 65 | |
| BC151073.1 | - | 2 - 3 | 1226 | 9087986 | 65 | |
| BC151075.1 | - | 2 - 3 | 1226 | 9087988 | 65 | |
More Information
Set Membership: |
|

Zfp608
Genomic Map