Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175751.4 | NP_786927.2 | 6 - 7 | 4665 | 95 | |
| GenBank mRNA | AK076178.1 | - | 2 - 3 | 1541 | 95 | |
| AK077030.1 | - | 5 - 6 | 3392 | 95 | ||
| AK129321.1 | - | 1 - 2 | 342 | 95 | ||
| BC059268.1 | - | 5 - 6 | 4331 | 95 | ||
| BC092219.1 | - | 6 - 7 | 4398 | 6830664 | 95 | |
| BC151073.1 | - | 5 - 6 | 4184 | 9087986 | 95 | |
| BC151075.1 | - | 5 - 6 | 4184 | 9087988 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp608
Genomic Map