Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175751.4 | NP_786927.2 | 8 - 9 | 5000 | 72 | |
| GenBank mRNA | AK077030.1 | - | 7 - 8 | 3731 | 72 | |
| BC059268.1 | - | 7 - 8 | 4666 | 72 | ||
| BC092219.1 | - | 8 - 9 | 4733 | 6830664 | 72 | |
| BC151073.1 | - | 7 - 8 | 4519 | 9087986 | 72 | |
| BC151075.1 | - | 7 - 8 | 4519 | 9087988 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp608
Genomic Map