Assay Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110273.1 | NP_001103743.1 | 2 - 3 | 826 | 90 | |
| NM_207651.3 | NP_997534.2 | 13 - 14 | 1966 | 90 | ||
| GenBank mRNA | AF366052.2 | - | 13 - 14 | 1958 | 90 | |
| AF367359.1 | - | 2 - 3 | 787 | 90 | ||
| AK032837.1 | - | 4 - 5 | 570 | 90 | ||
| BC150680.1 | - | 13 - 14 | 1874 | 9087591 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF367359.1, AK032837.1, NM_001110273.1 transcripts.
Set Membership: |
|

Slc14a2
Genomic Map