| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800011783.mm.1 |
NM_001110273.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011783.mm.1 |
NM_001110273.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011783.mm.1 |
NM_207651.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011783.mm.1 |
NM_207651.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011783.mm.1 |
TC1800001638.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011784.mm.1 |
NM_001110273.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011784.mm.1 |
NM_001110273.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011784.mm.1 |
NM_207651.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011784.mm.1 |
NM_207651.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011784.mm.1 |
TC1800001638.mm.1 |
MTA TC |