Assay Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030683.3 | NP_109608.1 | 2 - 3 | 375 | 95 | |
| NM_207651.3 | NP_997534.2 | 2 - 3 | 375 | 95 | ||
| GenBank mRNA | AF258602.1 | - | 2 - 3 | 276 | 95 | |
| AF366052.2 | - | 2 - 3 | 367 | 95 | ||
| BC150680.1 | - | 2 - 3 | 283 | 9087591 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF258602.1, AF366052.2, BC150680.1, NM_030683.3, NM_207651.3 transcripts.
Set Membership: |
|

Slc14a2
Genomic Map