Assay Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026470.3 | NP_080746.3 | 7 - 8 | 979 | 129 | |
| GenBank mRNA | AK005819.1 | - | 2 - 3 | 366 | 129 | |
| AK006861.1 | - | 2 - 3 | 351 | 129 | ||
| AK051574.1 | - | 7 - 8 | 980 | 129 | ||
| AK132771.1 | - | 7 - 8 | 953 | 129 | ||
| AK153098.1 | - | 7 - 8 | 856 | 129 | ||
| BC028429.1 | - | 2 - 3 | 379 | 129 | ||
| BC040024.1 | - | 2 - 3 | 379 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata6
Genomic Map