Assay Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164566.1 | NP_001158038.1 | 11 - 12 | 1419 | 89 | |
| GenBank mRNA | BC014764.1 | - | 11 - 12 | 1419 | 4217648 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Spats2l
Genomic Map