Assay Details
| Gene Symbol: Hsd3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146665.1 | - | 2 - 3 | 332 | 63 | |
| AK147114.1 | - | 3 - 4 | 522 | 63 | ||
| BC052659.1 | - | 2 - 3 | 263 | 30049155 | 63 | |
| M58567.1 | - | 2 - 3 | 276 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087805 | BC052659.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087805 | AK147114.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087805 | NM_008293.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2087805 | AK146665.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71673 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129447 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087805 | BC052659.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087805 | AK147114.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087805 | NM_008293.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2087805 | AK146665.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421418 | BC052659.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421418 | M58567.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421418 | AK147114.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421418 | NM_008293.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P421418 | AK146665.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22361179 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129447 | - | - |
| Whole Mouse Genome G4122A | A_51_P421418 | BC052659.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421418 | M58567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421418 | AK147114.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421418 | NM_008293.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P421418 | AK146665.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115141 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125803 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119882 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22361112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02423819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22361175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500568 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448453_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500565 | - | - |
| Mouse Genome 430 2.0 Array | 1448453_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448453_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15492_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15492_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300020045.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020045.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020041.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020041.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020047.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020047.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020044.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020044.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020048.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020048.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020040.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020040.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020042.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020042.mm.1 | TC0300002565.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300020046.mm.1 | NM_008293.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300020046.mm.1 | TC0300002565.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448453_at | - | - |
| Mouse Expression Set 430 | 1448453_PM_at | - | - |
| Murine Genome U74v2 Set | 161261_f_at | - | - |
| Murine Genome U74v2 Set | 103072_PM_at | - | - |
| Murine Genome U74v2 Set | 103072_at | - | - |
| Murine Genome U74v2 Set | 161261_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448453_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448453_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448453_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448453_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3060672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652602 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2652602 | - | - |
| mouse-6 v1.1 (Illumina) | scl21772.5_341-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21772.5_341-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2652602 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652602 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21772.5_341-S | - | - |

Hsd3b1
Genomic Map