Assay Details
| Gene Symbol: Mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145442.2 | NP_663417.1 | 5 - 6 | 732 | 99 | |
| GenBank mRNA | AK077125.1 | - | 5 - 6 | 653 | 99 | |
| AK154318.1 | - | 5 - 6 | 661 | 99 | ||
| AK171164.1 | - | 5 - 6 | 661 | 99 | ||
| BC002277.1 | - | 5 - 6 | 718 | 3496688 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mbip
Genomic Map