Assay Details
| Gene Symbol: Mbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025256.2 | NP_001020427.1 | 4 - 5 | 505 | 73 | |
| NM_001025259.2 | NP_001020430.1 | 3 - 4 | 427 | 73 | ||
| GenBank mRNA | AK005129.1 | - | 3 - 4 | 361 | 73 | |
| AK079621.1 | - | 4 - 5 | 461 | 73 | ||
| AK147966.1 | - | 3 - 4 | 375 | 73 | ||
| AK180082.1 | - | 2 - 3 | 82 | 73 | ||
| AK181508.1 | - | 2 - 3 | 82 | 73 | ||
| AK187476.1 | - | 2 - 3 | 126 | 73 | ||
| AK192745.1 | - | 2 - 3 | 82 | 73 | ||
| AK193213.1 | - | 2 - 3 | 82 | 73 | ||
| AK193221.1 | - | 2 - 3 | 82 | 73 | ||
| AK201106.1 | - | 2 - 3 | 82 | 73 | ||
| AK201437.1 | - | 2 - 3 | 73 | |||
| AK202471.1 | - | 2 - 3 | 82 | 73 | ||
| AK202837.1 | - | 2 - 3 | 82 | 73 | ||
| BB633793.1 | - | 4 - 5 | 461 | 73 | ||
| BY245551.1 | - | 3 - 4 | 427 | 73 | ||
| M15060.1 | - | 4 - 5 | 425 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mbp
Genomic Map