Assay Details
| Gene Symbol: 4921531P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045361.1 | 3 - 4 | 386 | 80 | ||
| GenBank mRNA | AK007141.1 | - | 3 - 4 | 387 | 80 | |
| AK014992.1 | - | 4 - 5 | 1198 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700095A13 gene | 1700095A13Rik | -212.1kb | Sense | Chr. 18 - 83076095 - 83080660 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31503 |
Gm31503 | 65.2kb | Sense | Chr. 18 - 83363538 - 83373741 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045216 | AK007141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045216 | AK014992.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13658295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13616093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13821028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14136347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31881340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431738_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431738_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431738_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800012095.mm.1 | NR_045361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012095.mm.1 | NR_045361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012095.mm.1 | TC1800001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800012096.mm.1 | NR_045361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012096.mm.1 | NR_045361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012096.mm.1 | TC1800001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800012089.mm.1 | NR_045361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012089.mm.1 | NR_045361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012089.mm.1 | TC1800001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800012090.mm.1 | NR_045361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012090.mm.1 | NR_045361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012090.mm.1 | TC1800001691.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800012088.mm.1 | NR_045361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012088.mm.1 | NR_045361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800012088.mm.1 | TC1800001691.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431738_at | - | - |
| Mouse Expression Set 430 | 1431738_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431738_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431738_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 592830 | - | - |

4921531P14Rik
Genomic Map