Assay Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177259.4 | NP_796233.2 | 9 - 10 | 1040 | 139 | |
| GenBank mRNA | BC138538.1 | - | 8 - 9 | 761 | 8861558 | 139 |
| BC138539.1 | - | 8 - 9 | 761 | 8861559 | 139 | |
| Y08379.1 | - | 9 - 10 | 988 | 139 | ||
More Information
Set Membership: |
|

Dab1
Genomic Map