Assay Details
| Gene Symbol: Lepr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122899.1 | NP_001116371.1 | 10 - 11 | 2148 | 116 | |
| NM_010704.2 | NP_034834.1 | 10 - 11 | 2148 | 116 | ||
| NM_146146.2 | NP_666258.2 | 10 - 11 | 2148 | 116 | ||
| GenBank mRNA | AK140785.1 | - | 10 - 11 | 1650 | 116 | |
| AK143989.1 | - | 10 - 11 | 1837 | 116 | ||
| AK155891.1 | - | 10 - 11 | 1688 | 116 | ||
| BC082551.1 | - | 10 - 11 | 1633 | 30621706 | 116 | |
| U42467.1 | - | 10 - 11 | 1656 | 116 | ||
| U46135.1 | - | 10 - 11 | 1656 | 116 | ||
| U49106.1 | - | 9 - 10 | 1596 | 116 | ||
| U49109.1 | - | 9 - 10 | 1596 | 116 | ||
| U49110.1 | - | 9 - 10 | 1596 | 116 | ||
| U52915.1 | - | 10 - 11 | 2110 | 116 | ||
| U58861.1 | - | 10 - 11 | 1678 | 116 | ||
| U58862.1 | - | 10 - 11 | 1688 | 116 | ||
| U58863.1 | - | 10 - 11 | 1678 | 116 | ||
| Y10296.1 | - | 9 - 10 | 1604 | 116 | ||
| Y10298.1 | - | 9 - 10 | 1604 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lepr
Genomic Map