Assay Details
| Gene Symbol: Lepr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122899.1 | NP_001116371.1 | 1 - 2 | 539 | 119 | |
| NM_010704.2 | NP_034834.1 | 1 - 2 | 539 | 119 | ||
| NM_146146.2 | NP_666258.2 | 1 - 2 | 539 | 119 | ||
| GenBank mRNA | AB519174.1 | - | 1 - 2 | 48 | 119 | |
| AB519175.1 | - | 1 - 2 | 57 | 119 | ||
| AK140785.1 | - | 1 - 2 | 41 | 119 | ||
| AK143989.1 | - | 1 - 2 | 228 | 119 | ||
| AK155891.1 | - | 1 - 2 | 79 | 119 | ||
| U52915.1 | - | 1 - 2 | 501 | 119 | ||
| U58861.1 | - | 1 - 2 | 69 | 119 | ||
| U58862.1 | - | 1 - 2 | 79 | 119 | ||
| U58863.1 | - | 1 - 2 | 69 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB519174.1, AB519175.1, AK140785.1, AK143989.1, NM_001122899.1, NM_010704.2, NM_146146.2, U52915.1, U58861.1, U58862.1, U58863.1 transcripts.
Set Membership: |
|

Lepr
Genomic Map