Assay Details
| Gene Symbol: Hook1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030014.2 | NP_084290.1 | 14 - 15 | 1505 | 118 | |
| GenBank mRNA | AF487912.1 | - | 14 - 15 | 1587 | 118 | |
| AK028714.1 | - | 14 - 15 | 1524 | 118 | ||
| AK043870.1 | - | 14 - 15 | 1512 | 118 | ||
| AK049897.1 | - | 14 - 15 | 1506 | 118 | ||
| BC061688.1 | - | 14 - 15 | 1505 | 5364426 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook1
Genomic Map