Assay Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293707.1 | NP_001280636.1 | 13 - 14 | 2072 | 72 | |
| NM_008847.3 | NP_032873.2 | 13 - 14 | 2069 | 72 | ||
| GenBank mRNA | AK098097.1 | - | 13 - 14 | 2026 | 72 | |
| AK150898.1 | - | 13 - 14 | 2029 | 72 | ||
| AK151984.1 | - | 13 - 14 | 2029 | 72 | ||
| AK158062.1 | - | 13 - 14 | 2014 | 72 | ||
| BC003763.1 | - | 10 - 11 | 1126 | 3591003 | 72 | |
| BC031774.1 | - | 13 - 14 | 1997 | 4503697 | 72 | |
| D86177.1 | - | 13 - 14 | 1859 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pip5k1a
Genomic Map