Assay Details
| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293715.1 | NP_001280644.1 | 5 - 6 | 1789 | 66 | |
| NM_001293716.1 | NP_001280645.1 | 3 - 4 | 807 | 66 | ||
| NM_175356.3 | NP_780565.2 | 4 - 5 | 1744 | 66 | ||
| GenBank mRNA | AK028974.1 | - | 4 - 5 | 1729 | 66 | |
| AK030704.1 | - | 4 - 5 | 1504 | 66 | ||
| AK048405.1 | - | 3 - 4 | 600 | 66 | ||
| AK053614.1 | - | 4 - 5 | 1739 | 66 | ||
| AK157918.1 | - | 4 - 5 | 1536 | 66 | ||
| BC079846.1 | - | 4 - 5 | 1744 | 5707855 | 66 | |
| BC113176.1 | - | 5 - 6 | 1644 | 40060773 | 66 | |
| BC113781.1 | - | 3 - 4 | 707 | 40060770 | 66 | |
| BC138457.1 | - | 4 - 5 | 1647 | 8861477 | 66 | |
| BC138458.1 | - | 4 - 5 | 1647 | 8861478 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pi4kb
Genomic Map