Assay Details
| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293715.1 | NP_001280644.1 | 6 - 7 | 1906 | 67 | |
| NM_001293716.1 | NP_001280645.1 | 4 - 5 | 924 | 67 | ||
| NM_175356.3 | NP_780565.2 | 5 - 6 | 1861 | 67 | ||
| GenBank mRNA | AK028974.1 | - | 5 - 6 | 1846 | 67 | |
| AK030704.1 | - | 5 - 6 | 1621 | 67 | ||
| AK053614.1 | - | 5 - 6 | 1856 | 67 | ||
| AK157918.1 | - | 5 - 6 | 1653 | 67 | ||
| AK172074.1 | - | 5 - 6 | 1854 | 67 | ||
| BC079846.1 | - | 5 - 6 | 1861 | 5707855 | 67 | |
| BC113176.1 | - | 6 - 7 | 1761 | 40060773 | 67 | |
| BC113781.1 | - | 4 - 5 | 824 | 40060770 | 67 | |
| BC138457.1 | - | 5 - 6 | 1764 | 8861477 | 67 | |
| BC138458.1 | - | 5 - 6 | 1764 | 8861478 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pi4kb
Genomic Map