Assay Details
| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293715.1 | NP_001280644.1 | 9 - 10 | 2401 | 82 | |
| NM_001293716.1 | NP_001280645.1 | 7 - 8 | 1419 | 82 | ||
| NM_175356.3 | NP_780565.2 | 8 - 9 | 2356 | 82 | ||
| GenBank mRNA | AK028974.1 | - | 8 - 9 | 2341 | 82 | |
| AK030704.1 | - | 8 - 9 | 2116 | 82 | ||
| AK048405.1 | - | 7 - 8 | 1212 | 82 | ||
| AK053614.1 | - | 8 - 9 | 2351 | 82 | ||
| AK157918.1 | - | 8 - 9 | 2148 | 82 | ||
| AK172074.1 | - | 8 - 9 | 2349 | 82 | ||
| BC059895.1 | - | 1 - 2 | 103 | 82 | ||
| BC079846.1 | - | 8 - 9 | 2356 | 5707855 | 82 | |
| BC113176.1 | - | 9 - 10 | 2256 | 40060773 | 82 | |
| BC113781.1 | - | 7 - 8 | 1319 | 40060770 | 82 | |
| BC138457.1 | - | 8 - 9 | 2259 | 8861477 | 82 | |
| BC138458.1 | - | 8 - 9 | 2259 | 8861478 | 82 | |
More Information
Set Membership: |
|

Pi4kb
Genomic Map