Assay Details
| Gene Symbol: Ino80d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081436.2 | NP_001074905.1 | 3 - 4 | 258 | 59 | |
| GenBank mRNA | AK049468.1 | - | 3 - 4 | 246 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049468.1, NM_001081436.2 transcripts.
Set Membership: |
|

Ino80d
Genomic Map