Assay Details
| Gene Symbol: Abi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198570.1 | NP_001185499.1 | 8 - 9 | 1218 | 89 | |
| NM_001198571.1 | NP_001185500.1 | 8 - 9 | 1119 | 89 | ||
| GenBank mRNA | BC079545.1 | - | 8 - 9 | 1004 | 6851145 | 89 |
| BC079646.1 | - | 8 - 9 | 1142 | 6850720 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abi2
Genomic Map