Assay Details
| Gene Symbol: Mppe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK172496.1 | - | 7 - 8 | 1072 | 82 | |
| BC045146.1 | - | 7 - 8 | 943 | 6306245 | 82 | |
| BC086456.1 | - | 7 - 8 | 990 | 4949311 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mppe1
Genomic Map