Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178793.4 | NP_848908.1 | 1 - 2 | 220 | 71 | |
| GenBank mRNA | AK028377.1 | - | 1 - 2 | 218 | 71 | |
| AK035153.1 | - | 1 - 2 | 215 | 71 | ||
| AK039742.1 | - | 1 - 2 | 150 | 71 | ||
| AK220435.1 | - | 1 - 2 | 190 | 71 | ||
| BC152322.1 | - | 1 - 2 | 189 | 40140631 | 71 | |
| CJ179960.1 | - | 1 - 2 | 220 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbe1
Genomic Map