Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003027.1 | - | 3 - 4 | 353 | 120 | |
| AK003411.1 | - | 3 - 4 | 334 | 120 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec11c| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003027.1 | - | 3 - 4 | 353 | 120 | |
| AK003411.1 | - | 3 - 4 | 334 | 120 | ||
Set Membership: |
|