Assay Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016792.4 | NP_058072.2 | 2 - 3 | 382 | 107 | |
| GenBank mRNA | AF052660.1 | - | 2 - 3 | 211 | 107 | |
| AK029807.1 | - | 2 - 3 | 383 | 107 | ||
| AK051698.1 | - | 2 - 3 | 357 | 107 | ||
| AK144211.1 | - | 2 - 3 | 339 | 107 | ||
| AK150326.1 | - | 2 - 3 | 345 | 107 | ||
| AK150887.1 | - | 2 - 3 | 351 | 107 | ||
| AK150983.1 | - | 2 - 3 | 351 | 107 | ||
| AK152101.1 | - | 2 - 3 | 351 | 107 | ||
| AK167954.1 | - | 2 - 3 | 353 | 107 | ||
| AK168712.1 | - | 2 - 3 | 346 | 107 | ||
| BC061123.1 | - | 2 - 3 | 409 | 6596299 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnl1
Genomic Map