Assay Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016792.4 | NP_058072.2 | 3 - 4 | 552 | 100 | |
| GenBank mRNA | AK029807.1 | - | 3 - 4 | 553 | 100 | |
| AK051698.1 | - | 3 - 4 | 527 | 100 | ||
| AK144211.1 | - | 3 - 4 | 509 | 100 | ||
| AK150326.1 | - | 3 - 4 | 515 | 100 | ||
| AK150887.1 | - | 3 - 4 | 521 | 100 | ||
| AK152101.1 | - | 3 - 4 | 521 | 100 | ||
| AK167954.1 | - | 3 - 4 | 523 | 100 | ||
| AK168712.1 | - | 3 - 4 | 516 | 100 | ||
| AK212711.1 | - | 1 - 2 | 134 | 100 | ||
| BC061123.1 | - | 3 - 4 | 579 | 6596299 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnl1
Genomic Map