Assay Details
| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025409.3 | NP_079685.1 | 1 - 2 | 194 | 87 | |
| GenBank mRNA | AK004281.1 | - | 1 - 2 | 179 | 87 | |
| AK007388.1 | - | 1 - 2 | 142 | 87 | ||
| AK007862.1 | - | 1 - 2 | 126 | 87 | ||
| AK009116.1 | - | 1 - 2 | 127 | 87 | ||
| AK210106.1 | - | 1 - 2 | 64 | 87 | ||
| BC021512.1 | - | 1 - 2 | 184 | 5351700 | 87 | |
| CX207811.1 | - | 1 - 2 | 183 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ier3ip1
Genomic Map