Assay Details
| Gene Symbol: Napg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028017.1 | NP_082293.1 | 7 - 8 | 480 | 92 | |
| GenBank mRNA | AK010275.1 | - | 7 - 8 | 533 | 92 | |
| AK090379.1 | - | 7 - 8 | 442 | 92 | ||
| AK154573.1 | - | 7 - 8 | 500 | 92 | ||
| AK164149.1 | - | 7 - 8 | 475 | 92 | ||
| AK167372.1 | - | 7 - 8 | 487 | 92 | ||
| BC026977.1 | - | 7 - 8 | 480 | 5362684 | 92 | |
More Information
Set Membership: |
|

Napg
Genomic Map