Assay Details
| Gene Symbol: Cxxc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028868.3 | NP_083144.1 | 11 - 12 | 1806 | 117 | |
| GenBank mRNA | AK010337.1 | - | 11 - 12 | 1807 | 117 | |
| AK083655.1 | - | 11 - 12 | 1811 | 117 | ||
| AK146688.1 | - | 11 - 12 | 1890 | 117 | ||
| AY099096.1 | - | 11 - 12 | 1778 | 117 | ||
| BC012261.1 | - | 7 - 8 | 1349 | 117 | ||
| BC030938.1 | - | 11 - 12 | 1638 | 3980339 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxxc1
Genomic Map