Assay Details
| Gene Symbol: Cxxc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028868.3 | NP_083144.1 | 13 - 14 | 1952 | 71 | |
| GenBank mRNA | AK010337.1 | - | 13 - 14 | 1953 | 71 | |
| AK083655.1 | - | 13 - 14 | 1957 | 71 | ||
| AK146688.1 | - | 13 - 14 | 2036 | 71 | ||
| AK179636.1 | - | 1 - 2 | 43 | 71 | ||
| AK184468.1 | - | 1 - 2 | 43 | 71 | ||
| AY099096.1 | - | 13 - 14 | 1924 | 71 | ||
| BC012261.1 | - | 9 - 10 | 1495 | 71 | ||
| BC030938.1 | - | 13 - 14 | 1784 | 3980339 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cxxc1
Genomic Map