Assay Details
| Gene Symbol: Acaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177470.3 | NP_803421.1 | 1 - 2 | 60 | 85 | |
| GenBank mRNA | AK050101.1 | - | 1 - 2 | 62 | 85 | |
| AK146007.1 | - | 1 - 2 | 75 | 85 | ||
| AK167567.1 | - | 1 - 2 | 61 | 85 | ||
| AK167715.1 | - | 1 - 2 | 58 | 85 | ||
| AK169359.1 | - | 1 - 2 | 61 | 85 | ||
| BC028901.1 | - | 1 - 2 | 55 | 4195488 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050101.1, AK146007.1, AK167567.1, AK167715.1, AK169359.1, BC028901.1, NM_177470.3 transcripts.
Set Membership: |
|

Acaa2
Genomic Map