Assay Details
| Gene Symbol: Pdgfrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146268.1 | NP_001139740.1 | 15 - 16 | 2615 | 83 | |
| NM_008809.2 | NP_032835.2 | 15 - 16 | 2612 | 83 | ||
| GenBank mRNA | AK134785.1 | - | 16 - 17 | 2637 | 83 | |
| AK143247.1 | - | 15 - 16 | 2595 | 83 | ||
| AK158145.1 | - | 9 - 10 | 1269 | 83 | ||
| BC055311.1 | - | 15 - 16 | 2627 | 30060666 | 83 | |
| DQ979390.1 | - | 9 - 10 | 1268 | 83 | ||
| X04367.1 | - | 15 - 16 | 2321 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfrb
Genomic Map